[5-(Trifluoromethyl)pyridin-2-yl]acetonitrile

Lieferant: Apollo Scientific

Synonyme: 2-(5-(Trifluoromethyl)pyridin-2-yl)acetonitrile

PC402036-1G
APOSPC402036-1GEA 63.8 EUR
APOSPC402036-1G
[5-(Trifluoromethyl)pyridin-2-yl]acetonitrile
[5-(Trifluoromethyl)pyridin-2-yl]acetonitrile
Formel: C₈H₅F₃N₂
Molecular Weight: 186,134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08690162
CAS-Nummer: 939793-18-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR