tert-Butyl 5-fluoro-3-iodo-1H-indole-1-carboxylate
Lieferant: Apollo Scientific
PC402039-250MG
APOSPC402039-250MGEA
128
EUR
APOSPC402039-250MG
tert-Butyl 5-fluoro-3-iodo-1H-indole-1-carboxylate
tert-Butyl 5-fluoro-3-iodo-1H-indole-1-carboxylate
Formel:
C₁₃H₁₃FINO₂ Molecular Weight: 361,151 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13183351 CAS-Nummer: 192189-13-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...