8-(Trifluoromethyl)-1,4-dioxaspiro[4.5]decan-8-ol

Lieferant: Apollo Scientific
Achtung

PC402041-250MG
APOSPC402041-250MGEA 561 EUR
APOSPC402041-250MG
8-(Trifluoromethyl)-1,4-dioxaspiro[4.5]decan-8-ol
8-(Trifluoromethyl)-1,4-dioxaspiro[4.5]decan-8-ol
Formel: C₉H₁₃F₃O₃
Molecular Weight: 226,193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16067897
CAS-Nummer: 1248081-37-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR