7-Fluoro-4-nitro-1H-benzimidazole

Lieferant: Apollo Scientific

PC402042-250MG
APOSPC402042-250MGEA 931 EUR
APOSPC402042-250MG
7-Fluoro-4-nitro-1H-benzimidazole
7-Fluoro-4-nitro-1H-benzimidazole
Formel: C₇H₄FN₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD26743819
CAS-Nummer: 18645-93-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR