5-Fluoro-6-hydroxy-2,3-dihydro-1H-isoindol-1-one
Lieferant: Apollo Scientific
Synonyme:
5-Fluoro-6-hydroxyisoindolin-1-one
PC402049-250MG
APOSPC402049-250MGEA
244
EUR
APOSPC402049-250MG
5-Fluoro-6-hydroxy-2,3-dihydro-1H-isoindol-1-one
5-Fluoro-6-hydroxy-2,3-dihydro-1H-isoindol-1-one
Formel:
C₈H₆FNO₂ Molecular Weight: 167,137 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12924344 CAS-Nummer: 1007455-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...