4-Fluoro-5-iodo-1H-indole

Lieferant: Apollo Scientific

PC402055-250MG
APOSPC402055-250MGEA 546 EUR
APOSPC402055-250MG
4-Fluoro-5-iodo-1H-indole
4-Fluoro-5-iodo-1H-indole
Formel: C₈H₅FIN
Lagertemperatur: Raumtemperatur
MDL: MFCD22628143
CAS-Nummer: 1240113-40-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR