4,6-Dichloro-7-fluoro-1,2-dihydro-3H-pyrrolo[3,4-c]pyridin-3-one

Lieferant: Apollo Scientific

PC402057-250MG
APOSPC402057-250MGEA 727 EUR
APOSPC402057-250MG
4,6-Dichloro-7-fluoro-1,2-dihydro-3H-pyrrolo[3,4-c]pyridin-3-one
4,6-Dichloro-7-fluoro-1,2-dihydro-3H-pyrrolo[3,4-c]pyridin-3-one
Formel: C₇H₃Cl₂FN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD20486199
CAS-Nummer: 1312693-69-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR