2,4-Dichloro-6-fluoroquinoline-3-carbonitrile

Lieferant: Apollo Scientific

PC402062-250MG
APOSPC402062-250MGEA 0 EUR
APOSPC402062-250MG
2,4-Dichloro-6-fluoroquinoline-3-carbonitrile
2,4-Dichloro-6-fluoroquinoline-3-carbonitrile
Formel: C₁₀H₃Cl₂FN₂
Molecular Weight: 241,049 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26142690
CAS-Nummer: 881313-89-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR