1-(Acetyloxy)ethyl 2-(2-fluorobiphenyl-4-yl)propanoate 98%

Lieferant: Apollo Scientific

Synonyme: 1-Acetoxyethyl 2-(2-fluoro-[1,1'-biphenyl]-4-yl)propanoate

PC402065-250MG
APOSPC402065-250MGEA 195 EUR
APOSPC402065-250MG
1-(Acetyloxy)ethyl 2-(2-fluorobiphenyl-4-yl)propanoate 98%
1-(Acetyloxy)ethyl 2-(2-fluorobiphenyl-4-yl)propanoate
Formel: C₁₉H₁₉FO₄
Molecular Weight: 330,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00900049
CAS-Nummer: 91503-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR