(S)-2-(4-Fluorophenyl)piperidine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

PC402100-100MG
APOSPC402100-100MGEA 168 EUR
APOSPC402100-100MG
(S)-2-(4-Fluorophenyl)piperidine hydrochloride 97%
(S)-2-(4-Fluorophenyl)piperidine hydrochloride
Formel: C₁₁H₁₄FN·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD12910636
CAS-Nummer: 1391431-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR