4-(3-(Trifluoromethyl)phenyl)-1,2,3,6-tetrahydropyridine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

PC402102-250MG
APOSPC402102-250MGEA 227 EUR
APOSPC402102-250MG
4-(3-(Trifluoromethyl)phenyl)-1,2,3,6-tetrahydropyridine hydrochloride 97%
4-(3-(Trifluoromethyl)phenyl)-1,2,3,6-tetrahydropyridine hydrochloride
Formel: C₁₂H₁₂F₃N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD22054946
CAS-Nummer: 1683-23-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR