3-Fluoro-5-(piperidin-4-yl)pyridine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

PC402103-100MG
APOSPC402103-100MGEA 132 EUR
APOSPC402103-100MG
3-Fluoro-5-(piperidin-4-yl)pyridine dihydrochloride 97%
3-Fluoro-5-(piperidin-4-yl)pyridine dihydrochloride
Formel: C₁₀H₁₃FN₂·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD29075592
CAS-Nummer: 1638968-09-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR