2-Fluoro-4-(piperidin-4-yl)phenol hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

PC402104-100MG
APOSPC402104-100MGEA 346 EUR
APOSPC402104-100MG
2-Fluoro-4-(piperidin-4-yl)phenol hydrochloride 97%
2-Fluoro-4-(piperidin-4-yl)phenol hydrochloride
Formel: C₁₁H₁₄FNO·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD29075599
CAS-Nummer: 1260871-71-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR