3-Fluoro-4-(piperidin-4-yl)pyridine dihydrochloride 95%

Lieferant: Apollo Scientific
Achtung

PC402105-250MG
APOSPC402105-250MGEA 676 EUR
APOSPC402105-250MG
3-Fluoro-4-(piperidin-4-yl)pyridine dihydrochloride 95%
3-Fluoro-4-(piperidin-4-yl)pyridine dihydrochloride
Formel: C₁₀H₁₃FN₂·2ClH
Molecular Weight: 253,144 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30099238
CAS-Nummer: 1260821-66-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR