2-Fluoro-4-(piperidin-4-yl)pyridine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

PC402106-100MG
APOSPC402106-100MGEA 0 EUR
APOSPC402106-100MG
2-Fluoro-4-(piperidin-4-yl)pyridine dihydrochloride 97%
2-Fluoro-4-(piperidin-4-yl)pyridine dihydrochloride
Formel: C₁₀H₁₃FN₂·2ClH
Molecular Weight: 253,144 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30099242
CAS-Nummer: 1138217-84-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR