(S)-2-(3,5-Difluorophenyl)piperidine hydrochloride 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC402107-100MG
APOSPC402107-100MGEA
168
EUR
APOSPC402107-100MG
(S)-2-(3,5-Difluorophenyl)piperidine hydrochloride 97%
(S)-2-(3,5-Difluorophenyl)piperidine hydrochloride
Formel:
C₁₁H₁₃F₂N·ClH Molecular Weight: 233,685 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26583092 CAS-Nummer: 1212946-48-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...