3-Fluoro-4-(piperidin-3-yl)pyridine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

PC402109-100MG
APOSPC402109-100MGEA 111 EUR
APOSPC402109-100MG
3-Fluoro-4-(piperidin-3-yl)pyridine dihydrochloride 97%
3-Fluoro-4-(piperidin-3-yl)pyridine dihydrochloride
Formel: C₁₀H₁₃FN₂·2ClH
Molecular Weight: 253,144 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30099271
CAS-Nummer: 1260645-80-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR