3-Fluoro-5-(piperidin-3-yl)pyridine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

PC402110-100MG
APOSPC402110-100MGEA 168 EUR
APOSPC402110-100MG
3-Fluoro-5-(piperidin-3-yl)pyridine dihydrochloride 97%
3-Fluoro-5-(piperidin-3-yl)pyridine dihydrochloride
Formel: C₁₀H₁₃FN₂·2ClH
Lagertemperatur: Kühlschrank
MDL: MFCD30099272
CAS-Nummer: 1260864-37-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR