5-Fluoro-4-methylindoline hydrochloride 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC402111-100MG
APOSPC402111-100MGEA
168
EUR
APOSPC402111-100MG
5-Fluoro-4-methylindoline hydrochloride 95%
5-Fluoro-4-methylindoline hydrochloride
Formel:
C₉H₁₀FN·ClH Molecular Weight: 187,642 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30730136 CAS-Nummer: 1388063-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...