3-Bromo-2-(chloromethyl)-6-fluoropyridine 97%

Lieferant: Apollo Scientific
Achtung

PC402113-100MG
APOSPC402113-100MGEA 168 EUR
APOSPC402113-100MG
3-Bromo-2-(chloromethyl)-6-fluoropyridine 97%
3-Bromo-2-(chloromethyl)-6-fluoropyridine
Formel: C₆H₄BrClFN
Molecular Weight: 224,458 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16608188
CAS-Nummer: 1227496-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR