3-(3,5-Difluorophenyl)piperidine 97%

Lieferant: Apollo Scientific
Achtung

PC402114-1G
APOSPC402114-1GEA 1030 EUR
APOSPC402114-1G
3-(3,5-Difluorophenyl)piperidine 97%
3-(3,5-Difluorophenyl)piperidine
Formel: C₁₁H₁₃F₂N
Molecular Weight: 197,224 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11047461
CAS-Nummer: 1044768-74-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR