(8-Fluoroquinolin-3-yl)boronic acid 95%

Lieferant: Apollo Scientific
Achtung

PC402116-1G
APOSPC402116-1GEA 243 EUR
APOSPC402116-1G
(8-Fluoroquinolin-3-yl)boronic acid 95%
(8-Fluoroquinolin-3-yl)boronic acid
Formel: C₉H₇BFNO₂
Molecular Weight: 190,967 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18382879
CAS-Nummer: 1207750-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR