trans-3-Fluoro-3-methylcyclobutane-1-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

PC402117-1G
APOSPC402117-1GEA 1860 EUR
APOSPC402117-1G
trans-3-Fluoro-3-methylcyclobutane-1-carboxylic acid 97%
trans-3-Fluoro-3-methylcyclobutane-1-carboxylic acid
Formel: C₆H₉FO₂
Lagertemperatur: Kühlschrank
MDL: MFCD28969665
CAS-Nummer: 1455037-43-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR