6-Fluorospiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-one 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC402119-1G
APOSPC402119-1GEA
121
EUR
APOSPC402119-1G
6-Fluorospiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-one 95%
6-Fluorospiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-one
Formel:
C₁₂H₁₃FN₂O₂ Molecular Weight: 236,2400 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD10566063 CAS-Nummer: 92926-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...