7-Fluor-4(3H)-chinazolinon 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Fluoro-4-hydroxyquinazoline, 7-Fluoro-4-quinazolinone, 7-Fluoro-4-quinazolinol

PC402122-5G
APOSPC402122-5GEA 89.9 EUR
APOSPC402122-5G
7-Fluor-4(3H)-chinazolinon 97%
7-Fluoroquinazolin-4(3H)-one
Formel: C₈H₅FN₂O
Molecular Weight: 164,1400 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD05664874
CAS-Nummer: 16499-57-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR