2-Bromo-1-(4-(fluoromethyl)phenyl)ethan-1-one 97%
Lieferant: Apollo Scientific
PC402128-1G
PC402128-500MG
APOSPC402128-1GEA
5240
EUR
APOSPC402128-1G
APOSPC402128-500MG
2-Bromo-1-(4-(fluoromethyl)phenyl)ethan-1-one 97%
2-Bromo-1-(4-(fluoromethyl)phenyl)ethan-1-one
Formel:
C₉H₈BrFO Molecular Weight: 231.062 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22492677 CAS-Nummer: 1628010-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...