2-Amino-7-(2,2,2-trifluoroethyl)-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidin-4(3H)-one 97%

Lieferant: Apollo Scientific

PC402129-1G
APOSPC402129-1GEA 2920 EUR
APOSPC402129-1G
2-Amino-7-(2,2,2-trifluoroethyl)-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidin-4(3H)-one 97%
2-Amino-7-(2,2,2-trifluoroethyl)-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidin-4(3H)-one
Formel: C₉H₁₁F₃N₄O
Molecular Weight: 248,205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629948
CAS-Nummer: 1782248-28-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR