(S)-2-(2,5-Difluorophenyl)pyrrolidine hydrochloride 97%

Lieferant: Apollo Scientific

PC402133-1G PC402133-5G
APOSPC402133-1GEA 300 EUR
APOSPC402133-1G APOSPC402133-5G
(S)-2-(2,5-Difluorophenyl)pyrrolidine hydrochloride 97%
(S)-2-(2,5-Difluorophenyl)pyrrolidine hydrochloride
Formel: C₁₀H₁₁F₂N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD08751479
CAS-Nummer: 1443624-23-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR