3-(4-Fluorophenyl)piperidine hydrochloride
Lieferant: Apollo Scientific
PC402142-1G
PC402142-250MG
APOSPC402142-1GEA
560
EUR
APOSPC402142-1G
APOSPC402142-250MG
3-(4-Fluorophenyl)piperidine hydrochloride
3-(4-Fluorophenyl)piperidine hydrochloride
Formel:
C₁₁H₁₅ClFN Lagertemperatur: Kühlschrank |
MDL:
MFCD02178931 CAS-Nummer: 1106940-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...