2-[3-(Trifluoromethyl)phenyl]propan-2-amine hydrochloride 97%

Lieferant: Apollo Scientific

Synonyme: 2-(3-(Trifluoromethyl)phenyl)propan-2-amine hydrochloride

PC403011-100MG PC403011-250MG PC403011-1G
APOSPC403011-100MGEA 111 EUR
APOSPC403011-100MG APOSPC403011-250MG APOSPC403011-1G
2-[3-(Trifluoromethyl)phenyl]propan-2-amine hydrochloride 97%
2-[3-(Trifluoromethyl)phenyl]propan-2-amine hydrochloride
Formel: C₁₀H₁₂F₃N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD14708148
CAS-Nummer: 153390-61-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR