2-[3-(Trifluoromethyl)phenyl]propan-2-amine hydrochloride 97%
Lieferant: Apollo Scientific
Synonyme:
2-(3-(Trifluoromethyl)phenyl)propan-2-amine hydrochloride
PC403011-100MG
PC403011-250MG
PC403011-1G
APOSPC403011-100MGEA
111
EUR
APOSPC403011-100MG
APOSPC403011-250MG
APOSPC403011-1G
2-[3-(Trifluoromethyl)phenyl]propan-2-amine hydrochloride 97%
2-[3-(Trifluoromethyl)phenyl]propan-2-amine hydrochloride
Formel:
C₁₀H₁₂F₃N·ClH Molecular Weight: 239.665 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14708148 CAS-Nummer: 153390-61-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...