3-(4-Fluorophenyl)-3-methylbutanoic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC403014-1G
APOSPC403014-1GEA
667
EUR
APOSPC403014-1G
3-(4-Fluorophenyl)-3-methylbutanoic acid
3-(4-Fluorophenyl)-3-methylbutanoic acid
Formel:
C₁₁H₁₃FO₂ Molecular Weight: 196,218 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11935534 CAS-Nummer: 339-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...