3-Fluoro-4-(oxetan-3-yloxy)aniline

Lieferant: Apollo Scientific
Achtung

PC403021-100MG
APOSPC403021-100MGEA 241 EUR
APOSPC403021-100MG
3-Fluoro-4-(oxetan-3-yloxy)aniline
3-Fluoro-4-(oxetan-3-yloxy)aniline
Formel: C₉H₁₀FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD20233094
CAS-Nummer: 1349716-61-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR