5-Amino-1-(2,2,2-trifluoroethyl)-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2,2,2-Trifluoroethyl)-1H-pyrazol-5-amine

PC403110-1G
APOSPC403110-1GEA 607 EUR
APOSPC403110-1G
5-Amino-1-(2,2,2-trifluoroethyl)-1H-pyrazole
5-Amino-1-(2,2,2-trifluoroethyl)-1H-pyrazole
Formel: C₅H₆F₃N₃
Molecular Weight: 165,116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16619962
CAS-Nummer: 899899-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR