(6-Fluoropyridin-3-yl)methylamine

Lieferant: Apollo Scientific

Synonyme: (6-Fluoropyridin-3-yl)methanamine

PC403115-1G PC403115-250MG PC403115-5G
APOSPC403115-1GEA 150 EUR
APOSPC403115-1G APOSPC403115-250MG APOSPC403115-5G
(6-Fluoropyridin-3-yl)methylamine
(6-Fluoropyridin-3-yl)methylamine
Formel: C₆H₇FN₂
Molecular Weight: 126.132 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100747
CAS-Nummer: 205744-17-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR