(6-Fluoropyridin-3-yl)methylamine
Lieferant: Apollo Scientific
Synonyme:
(6-Fluoropyridin-3-yl)methanamine
PC403115-1G
PC403115-250MG
PC403115-5G
APOSPC403115-1GEA
150
EUR
APOSPC403115-1G
APOSPC403115-250MG
APOSPC403115-5G
(6-Fluoropyridin-3-yl)methylamine
(6-Fluoropyridin-3-yl)methylamine
Formel:
C₆H₇FN₂ Molecular Weight: 126.132 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11100747 CAS-Nummer: 205744-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...