(5-Fluoropyridin-2-yl)methanamine 95%
Lieferant: Apollo Scientific
Synonyme:
2-(Aminomethyl)-5-fluoropyridine
PC403119-250MG
APOSPC403119-250MGEA
71.2
EUR
APOSPC403119-250MG
(5-Fluoropyridin-2-yl)methanamine 95%
(5-Fluoropyridin-2-yl)methanamine
Formel:
C₆H₇FN₂ Molecular Weight: 126,132 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD10697659 CAS-Nummer: 561297-96-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...