[1-(tert-Butoxycarbonyl)-4-(4-fluorophenyl)piperidin-4-yl]acetic acid

Lieferant: Apollo Scientific
Achtung

PC403198-1G
APOSPC403198-1GEA 800 EUR
APOSPC403198-1G
[1-(tert-Butoxycarbonyl)-4-(4-fluorophenyl)piperidin-4-yl]acetic acid
[1-(tert-Butoxycarbonyl)-4-(4-fluorophenyl)piperidin-4-yl]acetic acid
Formel: C₁₈H₂₄FNO₄
Molecular Weight: 337,386 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24849186
CAS-Nummer: 644981-80-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR