2-(Aminomethyl)-5-fluoropyridine dihydrochloride

Lieferant: Apollo Scientific
Achtung

PC403223-1G
APOSPC403223-1GEA 619 EUR
APOSPC403223-1G
2-(Aminomethyl)-5-fluoropyridine dihydrochloride
2-(Aminomethyl)-5-fluoropyridine dihydrochloride
Formel: C₆H₇FN₂·2ClH
Schmelzpunkt: 237…239 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16988819
CAS-Nummer: 859164-78-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR