2-Fluoro-5-methoxypyridine 98%

Lieferant: Apollo Scientific
Achtung

PC404001-10G PC404001-1G PC404001-5G
APOSPC404001-10GEA 374 EUR
APOSPC404001-10G APOSPC404001-1G APOSPC404001-5G
2-Fluoro-5-methoxypyridine 98%
2-Fluoro-5-methoxypyridine
Formel: C₆H₆FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD13185604
CAS-Nummer: 136888-79-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR