4-Amino-7-fluoroquinoline

Lieferant: Apollo Scientific

Synonyme: 4-Amino-7-fluoro-1-azanaphthalene, 7-Fluoroquinolin-4-amine

PC404502-1G
APOSPC404502-1GEA 1180 EUR
APOSPC404502-1G
4-Amino-7-fluoroquinoline
4-Amino-7-fluoroquinoline
Formel: C₉H₇FN₂
Molecular Weight: 162,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08063214
CAS-Nummer: 948293-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR