3-tert-Butoxycarbonylamino-3-(4-trifluoromethyl-phenyl)-propionic acid

Lieferant: Apollo Scientific

Synonyme: 3-((tert-Butoxycarbonyl)amino)-3-(4-(trifluoromethyl)phenyl)propanoic acid

PC404505-1G
APOSPC404505-1GEA 133 EUR
APOSPC404505-1G
3-tert-Butoxycarbonylamino-3-(4-trifluoromethyl-phenyl)-propionic acid
3-tert-Butoxycarbonylamino-3-(4-trifluoromethyl-phenyl)-propionic acid
Formel: C₁₅H₁₈F₃NO₄
Molecular Weight: 333.303 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571870
CAS-Nummer: 477849-00-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR