4-(3-Fluorobenzyl)piperidine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC404515-1G
APOSPC404515-1GEA 433 EUR
APOSPC404515-1G
4-(3-Fluorobenzyl)piperidine hydrochloride
4-(3-Fluorobenzyl)piperidine hydrochloride
Formel: C₁₂H₁₇ClFN
Molecular Weight: 229,723 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787475
CAS-Nummer: 193357-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR