4-(3-Trifluoromethyl-benzyl)-piperidine
Lieferant: Apollo Scientific
|
Achtung
|
PC404516-1G
APOSPC404516-1GEA
0
EUR
APOSPC404516-1G
4-(3-Trifluoromethyl-benzyl)-piperidine
4-(3-Trifluoromethyl-benzyl)-piperidine
Formel:
C₁₃H₁₆F₃N Lagertemperatur: Raumtemperatur |
MDL:
MFCD05863563 CAS-Nummer: 37581-28-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...