4-(5-Fluoro-2-methoxy-benzyl)-piperidine
Lieferant: Apollo Scientific
PC404518-1G
APOSPC404518-1GEA
467
EUR
APOSPC404518-1G
4-(5-Fluoro-2-methoxy-benzyl)-piperidine
4-(5-Fluoro-2-methoxy-benzyl)-piperidine
Formel:
C₁₃H₁₈FNO Molecular Weight: 223,287 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05863581 CAS-Nummer: 955314-86-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...