4-(3-Fluoro-4-methoxy-benzyl)-piperidine hydrochloride
Lieferant: Apollo Scientific
PC404519-1G
APOSPC404519-1GEA
433
EUR
APOSPC404519-1G
4-(3-Fluoro-4-methoxy-benzyl)-piperidine hydrochloride
4-(3-Fluoro-4-methoxy-benzyl)-piperidine hydrochloride
Formel:
C₁₃H₁₈FNO·ClH Molecular Weight: 259,747 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08751316 CAS-Nummer: 1171578-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...