4-(3-Fluoro-4-methyl-benzyl)-piperidine

Lieferant: Apollo Scientific

PC404520-1G
APOSPC404520-1GEA 0 EUR
APOSPC404520-1G
4-(3-Fluoro-4-methyl-benzyl)-piperidine
4-(3-Fluoro-4-methyl-benzyl)-piperidine
Formel: C₁₃H₁₈FN
Molecular Weight: 207.287 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863583
CAS-Nummer: 955287-58-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR