4-(4-Fluoro-2-methyl-benzyl)-piperidine

Lieferant: Apollo Scientific

PC404521-1G
APOSPC404521-1GEA 209 EUR
APOSPC404521-1G
4-(4-Fluoro-2-methyl-benzyl)-piperidine
4-(4-Fluoro-2-methyl-benzyl)-piperidine
Formel: C₁₃H₁₈FN
Molecular Weight: 207,287 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863584
CAS-Nummer: 955287-76-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR