4-(4-Fluoro-2-methyl-benzyl)-piperidine hydrochloride
Lieferant: Apollo Scientific
PC404522-1G
APOSPC404522-1GEA
185
EUR
APOSPC404522-1G
4-(4-Fluoro-2-methyl-benzyl)-piperidine hydrochloride
4-(4-Fluoro-2-methyl-benzyl)-piperidine hydrochloride
Formel:
C₁₃H₁₈FN·ClH Molecular Weight: 243,748 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09260414 CAS-Nummer: 1171960-54-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...