3-(4-Fluoro-benzyl)-piperidine

Lieferant: Apollo Scientific

PC404524-1G
APOSPC404524-1GEA 0 EUR
APOSPC404524-1G
3-(4-Fluoro-benzyl)-piperidine
3-(4-Fluoro-benzyl)-piperidine
Formel: C₁₂H₁₆FN
Molecular Weight: 193,261 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863601
CAS-Nummer: 382637-47-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR